missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat CNTF Protein

A cDNA sequence encoding the CNTF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10559-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15914248

  • 3915.00 € / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CNTF Protein
Research Use Only
Fully biologically active by its ability to phosphorylate STAT3 in several cells lines.
Rat
Untagged
Lyophilized from a concentrated (1 mg/ml) solution in water containing 0.025% NaHCO3.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
AFAEQTPLTL HRRDLSSRSIWLARKIRSDLTALMESYVKHQGLNKNINLDSVDGVPVASTDRWSEMTEAERLQENLQAYRTFQGMLTKLLEDQRVHFTPTEGDFHQAIHTLMLQVSAFAYQLEELMVLLEQKIPENEADGMPATVGDGGLFEKKLWGLKVLQELSQWTVRSIHDLRVISSHQMGISALESHYGAKDKQM
Greater than 99.0% as determined by:(a) Analysis by Gel Filtration. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers