missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat CXCL1/GRO alpha/KC/CINC-1 Protein

Product Code. 18607225 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18607225 5 μg 5µg
18613256 500 μg 500µg
18638625 1 mg 1mg
18655895 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18607225 Supplier R&D Systems™ Supplier No. NBP2352505UG
  • ONLINE EXCLUSIVE PRICE
    199.50 €
    175.35 € / 5µg
    Save 24.15 €
    12% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Recombinant Protein

CXCL1 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using rat neutrophils is in a concentration range of 10-100 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14095, P14095, P14095
For Use With (Application) SDS-PAGE, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl.
Gene ID (Entrez) 2919
Molecular Weight (g/mol) MolecularWeight-theroretical: 7.8 kDa
Quantity 5 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1 EU/μg of CXCL1/GRO alpha/KC/CINC-1 AS determined by LAL method.
Gene Alias CINC-1/CXCL1, FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1
Gene Symbol CXCL1
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Chemokines and Cytokines
Product Line Novus
Purity or Quality Grade >97 % pure by SDS-PAGE and HPLC
Protein CXCL1/GRO alpha/KC/CINC-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.