missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Guanidinoacetate N-methyltransferase Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Guanidinoacetate N-methyltransferase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6068-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929749

  • 735.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Guanidinoacetate N-methyltransferase Protein
Research Use Only
Gamt
Recombinant Protein
E. coli
MSSSAASPLF APGEDCGPAW RAAPAAYDTS DTHLQILGKP VMERWETPYM HSLAAAAASR GGRVLEVGFG MAIAASRVQQ APIKEHWIIE CNDGVFQRLQ NWALKQPHKV VPLKGLWEEE APTLPDGHFD GILYDTYPLS EETWHTHQFN FIKTHAFRLL KPGGILTYCN LTSWGELMKS KYTDITAMFE ETQVPALLEA GFQRENICTE VMALVPPADC RYYAFPQMIT PLVTKH
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers