missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Guanidinoacetate N-methyltransferase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Guanidinoacetate N-methyltransferase, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Name | Guanidinoacetate N-methyltransferase Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | Gamt |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929749
|
enQuireBio™
QP6068-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949749
|
enQuireBio™
QP6068-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939749
|
enQuireBio™
QP6068-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15959749
|
enQuireBio™
QP6068-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969749
|
enQuireBio™
QP6068-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15979749
|
enQuireBio™
QP6068-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Guanidinoacetate N-methyltransferase Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
Gamt | |
Recombinant Protein | |
E. coli | |
MSSSAASPLF APGEDCGPAW RAAPAAYDTS DTHLQILGKP VMERWETPYM HSLAAAAASR GGRVLEVGFG MAIAASRVQQ APIKEHWIIE CNDGVFQRLQ NWALKQPHKV VPLKGLWEEE APTLPDGHFD GILYDTYPLS EETWHTHQFN FIKTHAFRLL KPGGILTYCN LTSWGELMKS KYTDITAMFE ETQVPALLEA GFQRENICTE VMALVPPADC RYYAFPQMIT PLVTKH | |
Greater than 90% as determined by SDS-PAGE. |