missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL 10 Protein

A cDNA sequence encoding the IL 10 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10688-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954658

  • 131.00 € / 2µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

25325
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 as determined by the dose dependent inhibition of MC/9 proliferation is less than 10ng/ml concentration corresponding to a Specific Activity of 100,000IU/mg.
Rat
Untagged
The protein was lyophilized containing 20mM Tris-HCl pH 8.0 and 100mM NaCl.
IL 10 Protein
Research Use Only
Il10
Recombinant Protein
E. coli
SKGHSIRGDN NCTHFPVSQT HMLRELRAAF SQVKTFFQKK DQLDNILLTD SLLQDFKGYL GCQALSEMIK FYLVEVMPQA ENHGPEIKEH LNSLGEKLKT LWIQLRRCHR FLPCENKSKA VEQVKNDFNK LQDKGVYKAM NEFDIFINCI EAYVTLKMKN
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers