missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 10 Protein
A cDNA sequence encoding the IL 10 was constructed and used to recombinantly synthesize the protein.
131.00 € - 6790.00 €
Specifications
Gene ID (Entrez) | 25325 |
---|---|
Name | IL 10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Il10 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954658
|
enQuireBio™
QP10688-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934658
|
enQuireBio™
QP10688-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944658
|
enQuireBio™
QP10688-1MG |
1 mg |
6790.00 €
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
25325 | |
Research Use Only | |
Il10 | |
Recombinant Protein | |
E. coli | |
SKGHSIRGDN NCTHFPVSQT HMLRELRAAF SQVKTFFQKK DQLDNILLTD SLLQDFKGYL GCQALSEMIK FYLVEVMPQA ENHGPEIKEH LNSLGEKLKT LWIQLRRCHR FLPCENKSKA VEQVKNDFNK LQDKGVYKAM NEFDIFINCI EAYVTLKMKN | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 10 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by the dose dependent inhibition of MC/9 proliferation is less than 10ng/ml concentration corresponding to a Specific Activity of 100,000IU/mg. | |
Rat | |
Untagged | |
The protein was lyophilized containing 20mM Tris-HCl pH 8.0 and 100mM NaCl. |