missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL 33 Protein

A cDNA sequence encoding the IL 33 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10715-1mg


Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954948

  • 6790.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

361749
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
SIQGTSLLTE SCALSTYNDQ SVSFVLENGC YVINVEDCGK NQEKDKVLLR YYESSFPAQS GDGVDGKKLM VNMSPIKDTD IWLNANDKDY SVELQKGDVS PPDQAFFVLH KKSSDFVSFE CKNLPGTYIG VKDNQLALVE ENDESCNNIM FKLSKM
Greater than 95.0% as determined by SDS-PAGE.
IL 33 Protein
Research Use Only
Il33
Rat
Untagged
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers