missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 33 Protein
A cDNA sequence encoding the IL 33 was constructed and used to recombinantly synthesize the protein.
131.00 € - 6790.00 €
Specifications
Gene ID (Entrez) | 361749 |
---|---|
Name | IL 33 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Il33 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964948
|
enQuireBio™
QP10715-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944948
|
enQuireBio™
QP10715-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954948
|
enQuireBio™
QP10715-1MG |
1 mg |
6790.00 €
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
361749 | |
Research Use Only | |
Il33 | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
IL 33 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
SIQGTSLLTE SCALSTYNDQ SVSFVLENGC YVINVEDCGK NQEKDKVLLR YYESSFPAQS GDGVDGKKLM VNMSPIKDTD IWLNANDKDY SVELQKGDVS PPDQAFFVLH KKSSDFVSFE CKNLPGTYIG VKDNQLALVE ENDESCNNIM FKLSKM | |
Greater than 95.0% as determined by SDS-PAGE. |