missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 33 Protein
A cDNA sequence encoding the IL 33 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10715-2ug
Additional Details : Weight : 0.01000kg
Specifications
361749 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
SIQGTSLLTE SCALSTYNDQ SVSFVLENGC YVINVEDCGK NQEKDKVLLR YYESSFPAQS GDGVDGKKLM VNMSPIKDTD IWLNANDKDY SVELQKGDVS PPDQAFFVLH KKSSDFVSFE CKNLPGTYIG VKDNQLALVE ENDESCNNIM FKLSKM | |
Greater than 95.0% as determined by SDS-PAGE. |
IL 33 Protein | |
Research Use Only | |
Il33 | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |