missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 5 Protein
A cDNA sequence encoding the IL 5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10718-10ug
Additional Details : Weight : 0.01000kg
Specifications
24497 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 range=0.3-1.0 ng/ml as determined by the dose-dependant stimulation of the proliferation of BCL-1 cells. | |
Rat | |
Untagged | |
Lyophilized from a concentrated (1 mg/ml) solution in water containing no additives. |
IL 5 Protein | |
Research Use Only | |
Il5 | |
Recombinant Protein | |
E. coli | |
MRMLLCLNVL TLSCVWAIAM EIPMSTVVKE TLIQLSTHRA LLTSNETMRL PVPTHKNHQL CIGEIFQGLD ILKNQTVRGG TVEILFQNLS LIKKYIDGQK EKCGEERRKT RHFLDYLQEF LGVMSTEWAM EV | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |