missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Leptin Protein
Recombinant Protein
Brand: enQuireBio™ QP5507-5mg
Additional Details : Weight : 0.01000kg
Specifications
NP_037208.1 | |
Leptin Protein | |
5 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MVPIHKVQDD TKTLIKTIVT RINDISHTQS VSARQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQILTSLP SQNVLQIAHD LENLRDLLHL LAFSKSCSLP QTRGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDLSPEC |
25608 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
LEP | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |