missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Leptin Protein
Recombinant Protein
107.00 € - 1245.00 €
Specifications
Accession Number | NP_037208.1 |
---|---|
Gene ID (Entrez) | 25608 |
Name | Leptin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946279
|
enQuireBio™
QP5507-200UG |
200 μg |
107.00 €
200µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936279
|
enQuireBio™
QP5507-1MG |
1 mg |
252.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956279
|
enQuireBio™
QP5507-5MG |
5 mg |
690.00 €
5mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926279
|
enQuireBio™
QP5507-10MG |
10 mg |
1245.00 €
10mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_037208.1 | |
Leptin Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MVPIHKVQDD TKTLIKTIVT RINDISHTQS VSARQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQILTSLP SQNVLQIAHD LENLRDLLHL LAFSKSCSLP QTRGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDLSPEC | |
Greater than 95% as determined by SDS-PAGE. |
25608 | |
Research Use Only | |
LEP | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |