missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Leptin Protein

Recombinant Protein

Brand:  enQuireBio™ QP5507-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936279

  • 252.00 € / 1mg
Estimated Shipment: 05-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_037208.1
Leptin Protein
Research Use Only
LEP
Rat
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
25608
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MVPIHKVQDD TKTLIKTIVT RINDISHTQS VSARQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQILTSLP SQNVLQIAHD LENLRDLLHL LAFSKSCSLP QTRGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDLSPEC
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers