missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Minpp1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Minpp1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6360-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042740

  • 3260.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_062136.1
Minpp1 Protein
Research Use Only
Minpp1
Recombinant Protein
Yeast
SLPGRGDPVA SVLSPYFGTK TRYEDVNPWL LGDPVAPRRD PELLAGTCTP VQLVALIRHG TRYPTTKQIR KLRQLQGLLQ TRESVDGGSR VAAALDQWPL WYDDWMDGQL VEKGRQDMRQ LALRLAALFP DLFCRENYGR LRLITSSKHR CVDSSAAFLQ GLWQHYHPGL PPPDVSDMEC DPPRVNDKLM RFFDHCEKFL TEVERNATAL YHVEAFKTGP EMQTVLKKVA ATLQVPVNNL NADLIQVAFF TCSFDLAIQG VHSPWCDVFD VDDAKVLEYL NDLKQYWKRS YGYAINSRSS CNLFQDIFLH LDKAVEQKQR SQPVSSSVIL QFGHAETLLP LLSLMGYFKD KEPLTAYNFE EQVHREFRSG HIVPYASNLI FVLYHCEDAQ TPQEKFQIQM LLNEKVLPLA HSQKTVALYE DLKNHYQDIL QSCQTSKECN LPKVNITSDE L
Greater than 90% as determined by SDS-PAGE.
29688
1 mg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers