missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Minpp1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Minpp1, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_062136.1 |
---|---|
Gene ID (Entrez) | 29688 |
Name | Minpp1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022740
|
enQuireBio™
QP6360-YE-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042740
|
enQuireBio™
QP6360-YE-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032740
|
enQuireBio™
QP6360-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052740
|
enQuireBio™
QP6360-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062740
|
enQuireBio™
QP6360-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072740
|
enQuireBio™
QP6360-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_062136.1 | |
Minpp1 Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
29688 | |
Research Use Only | |
Minpp1 | |
Recombinant Protein | |
Yeast | |
SLPGRGDPVA SVLSPYFGTK TRYEDVNPWL LGDPVAPRRD PELLAGTCTP VQLVALIRHG TRYPTTKQIR KLRQLQGLLQ TRESVDGGSR VAAALDQWPL WYDDWMDGQL VEKGRQDMRQ LALRLAALFP DLFCRENYGR LRLITSSKHR CVDSSAAFLQ GLWQHYHPGL PPPDVSDMEC DPPRVNDKLM RFFDHCEKFL TEVERNATAL YHVEAFKTGP EMQTVLKKVA ATLQVPVNNL NADLIQVAFF TCSFDLAIQG VHSPWCDVFD VDDAKVLEYL NDLKQYWKRS YGYAINSRSS CNLFQDIFLH LDKAVEQKQR SQPVSSSVIL QFGHAETLLP LLSLMGYFKD KEPLTAYNFE EQVHREFRSG HIVPYASNLI FVLYHCEDAQ TPQEKFQIQM LLNEKVLPLA HSQKTVALYE DLKNHYQDIL QSCQTSKECN LPKVNITSDE L | |
Greater than 90% as determined by SDS-PAGE. |