missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Nitric oxide synthase, endothelial Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Nitric oxide synthase, endothelial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8865-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036891

  • 3375.00 € / 1mg
Estimated Shipment: 05-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_068610.1
Nitric oxide synthase, endothelial Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
24600
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Nos3
Recombinant Protein
E. coli
ATILYGSETG RAQSYAQQLG RLFRKAFDPR VLCMDEYDVV SLEHEALVLV VTSTFGNGDP PENGESFAAA LMEMSGPYNS SPRPEQHKSY KIRFNSVSCS DPLVSSWRRK RKESSNTDSA GALGTLRFCV FGLGSRAYPH FCAFARAVDT RLEELGGERL LQLGQGDELC GQ
Videos
SDS
Documents
Certificates
Special Offers

Special Offers