missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat S100A9 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) S100A9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6644-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044770

  • 3260.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_446039.1
S100A9 Protein
Research Use Only
S100A9
Recombinant Protein
E. coli
AAKTGSQLER SISTIINVFH QYSRKYGHPD TLNKAEFKEM VNKDLPNFLK REKRNENLLR DIMEDLDTNQ DNQLSFEECM MLMGKLIFAC HEKLHENNPR GHDHSHGKGC GK
Greater than 80% as determined by SDS-PAGE.
94195
1 mg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers