missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SCF / C-kit ligand Protein
Recombinant Protein
Brand: enQuireBio™ QP5517-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_068615.1 | |
SCF / C-kit ligand Protein | |
Research Use Only | |
KITLG | |
Recombinant Protein | |
E. coli | |
MQEICRNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV THLSVSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVAC MEENAPKNVK ESLKKPETRN FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA | |
Greater than 95% as determined by SDS-PAGE. |
60427 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat SCF / C-kit ligand induces TF-1 cell proliferation (typical ED50 is ? 15 ng/ml). | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |