missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SCF / C-kit ligand Protein
Recombinant Protein
107.00 € - 4060.00 €
Specifications
Accession Number | NP_068615.1 |
---|---|
Gene ID (Entrez) | 60427 |
Name | SCF / C-kit ligand Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15996319
|
enQuireBio™
QP5517-2UG |
2 μg |
107.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976319
|
enQuireBio™
QP5517-10UG |
10 μg |
252.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966319
|
enQuireBio™
QP5517-100UG |
100 μg |
630.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906329
|
enQuireBio™
QP5517-500UG |
500 μg |
2490.00 €
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986319
|
enQuireBio™
QP5517-1MG |
1 mg |
4060.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_068615.1 | |
SCF / C-kit ligand Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat SCF / C-kit ligand induces TF-1 cell proliferation (typical ED50 is ? 15 ng/ml). | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
60427 | |
Research Use Only | |
KITLG | |
Recombinant Protein | |
E. coli | |
MQEICRNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV THLSVSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVAC MEENAPKNVK ESLKKPETRN FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA | |
Greater than 95% as determined by SDS-PAGE. |