missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rhesus Macaque IL-4 Protein

Product Code. 18620746 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18620746 500 μg 500µg
18602286 5 μg 5µg
18683426 1 mg 1mg
18662496 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18620746 Supplier R&D Systems™ Supplier No. NBP235309500UG
  • ONLINE EXCLUSIVE PRICE
    5325.27 €
    4731.30 € / 500µg
    Save 593.97 €
    11% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Recombinant Protein

IL4 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 1.0 ng/mL, corresponding to a specific activity of > 1.0 x 10^6 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51492, P51492, P51492
For Use With (Application) SDS-PAGE, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in 2 x PBS, pH 7.4.
Gene ID (Entrez) 3565
Molecular Weight (g/mol) MolecularWeight-theroretical: 14.9 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 1 EU/μg of IL-4 AS determined by LAL method.
Gene Alias B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4 B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Gene Symbol IL4
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Biologically Active Proteins
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein IL-4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.