missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus 30 kDa neutral phosphatase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Oxygen-insensitive NADPH nitroreductase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7091-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017360

  • 1025.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

S. aureus 30 kDa neutral phosphatase Protein
Research Use Only
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
100 μg
Not determined.
Recombinant Protein
E. coli
KSSAEVQQTQ QASIPASQKA NLGNQNNIMX VAXYQ
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers