missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus 30 kDa neutral phosphatase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Oxygen-insensitive NADPH nitroreductase, was expressed in the hosts and tags indicated.
421.00 € - 3590.00 €
Specifications
Name | S. aureus 30 kDa neutral phosphatase Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Protein Length | Full Length |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027360
|
enQuireBio™
QP7091-10UG |
10 μg |
421.00 €
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067360
|
enQuireBio™
QP7091-50UG |
50 μg |
605.00 €
50µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017360
|
enQuireBio™
QP7091-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047360
|
enQuireBio™
QP7091-200UG |
200 μg |
1690.00 €
200µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057360
|
enQuireBio™
QP7091-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037360
|
enQuireBio™
QP7091-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
S. aureus 30 kDa neutral phosphatase Protein | |
Not determined. | |
Recombinant Protein | |
E. coli | |
KSSAEVQQTQ QASIPASQKA NLGNQNNIMX VAXYQ | |
Greater than 90% as determined by SDS-PAGE. |
Research Use Only | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |