missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus esxA Protein
A DNA sequence encoding the Staphylococcus aureus (strain MSSA476) esxA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7776-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
WP_001240826.1 | |
S. aureus esxA Protein | |
1 mg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
31213083 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
esxA | |
Recombinant Protein | |
E. coli | |
MAMIKMSPEE IRAKSQSYGQ GSDQIRQILS DLTRAQGEIA ANWEGQAFSR FEEQFQQLSP KVEKFAQLLE EIKQQLNSTA DAVQEQDQQL SNNFGLQ |