Learn More
enQuireBio™ Recombinant S. cerevisiae Homocysteine S-methyltransferase 1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Homocysteine S-methyltransferase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8787-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_013038.1 | |
S. cerevisiae Homocysteine S-methyltransferase 1 Protein | |
1 mg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |
850664 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
MHT1 | |
Recombinant Protein | |
E. coli | |
MKRIPIKELI VEHPGKVLIL DGGQGTELEN RGININSPVW SAAPFTSESF WEPSSQERKV VEEMYRDFMI AGANILMTIT YQANFQSISE NTSIKTLAAY KRFLDKIVSF TREFIGEERY LIGSIGPWAA HVSCEYTGDY GPHPENIDYY GFFKPQLENF NQNRDIDLIG FETIPNFHEL KAILSWDEDI ISKPFYIGLS VDDNSLLRDG TTLEEISVHI KGLGNKINKN LLLMGVNCVS FNQSALILKM LHEHLPGMPL LVYPNSGEIY NPKEKTWHRP TNKLDDWETT VKKFVDNGAR IIGGCCRTSP KDIAEIASAV DKYS |