missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Neutral trehalase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Neutral trehalase, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_010284.1 |
---|---|
Gene ID (Entrez) | 851564 |
Name | S. cerevisiae Neutral trehalase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017191
|
enQuireBio™
QP8922-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037191
|
enQuireBio™
QP8922-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027191
|
enQuireBio™
QP8922-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047191
|
enQuireBio™
QP8922-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057191
|
enQuireBio™
QP8922-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067191
|
enQuireBio™
QP8922-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_010284.1 | |
S. cerevisiae Neutral trehalase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |
851564 | |
Research Use Only | |
NTH1 | |
Recombinant Protein | |
E. coli | |
YMWDDESGFF FDYNTKIKHR TSYESATTFW ALWAGLATKE QAQKMVEKAL PKLEMLGGLA ACTERSRGPI SISRPIRQWD YPFGWAPHQI LAWEGLRSYG YLTVTNRLAY RWLFMMTKAF VDYNGIVVEK YDVTRGTDPH RVEAEYGNQG ADFKGAATEG FGWVNASYIL GLKYMNSHAR RALGACIPPI SFFSSLRPQE RNLYGL | |
Greater than 90% as determined by SDS-PAGE. |