missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant SARS SARS MERS Protein
A cDNA sequence encoding the SARS MERS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13419-500ug
Additional Details : Weight : 0.01000kg
Specifications
SARS SARS MERS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
EAKPSGSVVEQAEGVECDFSPLLSGTPPQVYNFKRLVFTNCNYNLTKLLSLFSVNDFTCSQISPAAIASNCYSSLILDYFSYPLSMKSDLSVSSAGPISQFNYKQSFSNPTCLILATVPHNLTTITKPLKYSYINKCSRLLSDDRTEVPQLVNANQYSPCVSIVPSTVWEDGDYYRKQLSPLEGGGWLVASGSTVAMTEQLQMGFGITVQYGTDTNSVCPKLEFANDTKIASQLGNCVEYHHHHHH | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
SARS | |
His | |
SARS MERS protein solution is supplied in PBS, 25mM arginine and 0.05% sodium azide. |