missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Soybean Soybean P34 GST Protein
A cDNA sequence encoding the Soybean P34 GST was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13564-1mg
Additional Details : Weight : 0.01000kg
Specifications
P34 GST Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, pH 8.0, 60mM NaCl, 10mM glutathione and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Soybean | |
GST | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |