missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Unstained Protein Ladder (14-98 kDa)

Product Code. 18656251 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18656251 100 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18656251 Supplier R&D Systems™ Supplier No. NBP311649
  • ONLINE EXCLUSIVE PRICE
    247.59 €
    228.90 € / 1 sets
    Save 18.69 €
    7% Off
View all promotions
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Recommended use in 12% SDS-PAGE
Quantity 100 Tests
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.