missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Xenopus Histone H1.0-A Protein
A DNA sequence encoding the Xenopus laevis (African clawed frog) Histone H1.0-A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7151-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001082697.1 | |
Histone H1.0-A Protein | |
Research Use Only | |
h1f0-a | |
Recombinant Protein | |
E. coli | |
MTENSAPAAK PRRSKASKKS TDHPKYSDMI LDAVQAEKSR SGSSRQSIQK YIKNNYTVGE NADSQIKLSI KRLVTSGTLK QTKGVGASGS FRLAKADEVK KPAKKPKKEI KKAVSPKKAA KPKKAAKSPA KAKKPKVAEK KVKKAPKKKP APSPRKAKKT KTVRAKPVWA SKAKKAKPSK PKAKASPKKS GRKK | |
Greater than 90% as determined by SDS-PAGE. |
398662 | |
100 μg | |
Not determined. | |
Full Length | |
Xenopus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |