missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Xenopus Histone H1.0-A Protein
A DNA sequence encoding the Xenopus laevis (African clawed frog) Histone H1.0-A, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_001082697.1 |
---|---|
Gene ID (Entrez) | 398662 |
Name | Histone H1.0-A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017490
|
enQuireBio™
QP7151-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037490
|
enQuireBio™
QP7151-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027490
|
enQuireBio™
QP7151-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047490
|
enQuireBio™
QP7151-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057490
|
enQuireBio™
QP7151-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067490
|
enQuireBio™
QP7151-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001082697.1 | |
Histone H1.0-A Protein | |
Not determined. | |
Full Length | |
Xenopus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
398662 | |
Research Use Only | |
h1f0-a | |
Recombinant Protein | |
E. coli | |
MTENSAPAAK PRRSKASKKS TDHPKYSDMI LDAVQAEKSR SGSSRQSIQK YIKNNYTVGE NADSQIKLSI KRLVTSGTLK QTKGVGASGS FRLAKADEVK KPAKKPKKEI KKAVSPKKAA KPKKAAKSPA KAKKPKVAEK KVKKAPKKKP APSPRKAKKT KTVRAKPVWA SKAKKAKPSK PKAKASPKKS GRKK | |
Greater than 90% as determined by SDS-PAGE. |