missing translation for 'onlineSavingsMsg'
Learn More

Redox Standard, 250mV, Reagecon™
BIOPHARMA

Product Code. 11803570 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11803570 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11803570 Supplier Reagecon Supplier No. RS250
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Controls the functionality and environmental conditions of redox sensing and reference electrodes. Redox Standard, 250mV, Reagecon™ is a ready to use solution of iron salts that is certified with a verifiable accuracy of ±5mV (with platinum electrode and Ag/AgCl reference solution (3M KCl).

Redox (reduction–oxidation) reactions cause an electron transfer process that can be measured using a Redox or ORP (oxidation-reduction potential) electrode. Redox is an absolute measurement expressed in millivolts. During its working life, a Redox electrode undergoes no change of zero point or slope. Therefore, redox electrodes do not require calibration. A control standard controls the functionality of the sensing and reference electrode, and also controls the analyst's technique, environmental conditions and the operation of the measurement meter.

Specifications

Packaging 500mL
CAS 13746-66-2, 13943-58-3
Vapor Pressure 23 hPa
Conductivity 250mV ±5
Physical Form Liquid
Molecular Formula C6FeK3N6
MDL Number MFCD00011392
Boiling Point 100°C
For Use With (Equipment) Redox Electrode
Solubility Completely miscible
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
IUPAC Name tripotassium hexacyanoirontris(ylium)
Molecular Weight (g/mol) 329.25
PubChem CID 26250
Quantity 500 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.