Learn More
Reelin Antibody (CL5324) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61414V
Product Code. 30289411
  • 737.00 € / 0.10mL
Description
Reelin Monoclonal antibody specifically detects Reelin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reelin
Monoclonal
DyLight 405
EC 3.4.21, EC 3.4.21.-, reelin, RLPRO1598
This antibody was developed using a recombinant protein derived from P78509, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Mouse, Rat
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL5324
50mM Sodium Borate
Mouse
Protein A purified
RUO
5649
Store at 4⪚C in the dark.
IgG1