missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reelin Antibody (CL5324) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61414PE
This item is not returnable.
View return policy
Description
Reelin Monoclonal antibody specifically detects Reelin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Reelin | |
| Monoclonal | |
| PE | |
| EC 3.4.21, EC 3.4.21.-, reelin, RLPRO1598 | |
| This antibody was developed using a recombinant protein derived from P78509, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL5324 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5649 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction