missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ REEP2 Recombinant Rabbit Monoclonal Antibody (K01_1X72)
GREENER_CHOICE

Product Code. p-200114931
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829950 100 μL
30830438 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829950 Supplier Invitrogen™ Supplier No. MA563431

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DALPLQRPDG RLRP.

Mammalian odorant receptors require accessory proteins such as RTP1 and RTP2 for functional cell surface expression. Receptor expression-enhancing protein (REEP) family members are transmembrane proteins which interact with odorant receptor proteins and may enhance the odorant receptor responses to odorants. Recently studies have shown other chemosensory receptors such as bitter taste receptors are also influenced by RTP and REEP family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REEP2
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1X72
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene REEP2
Gene Accession No. Q9BRK0
Gene Alias C2orf19; C5orf19; LOC532432 protein; LOC682105; receptor accessory protein 2; receptor expression-enhancing protein 2; Reep2; SGC32445; SPG72; Yip2d
Gene Symbols REEP2
Host Species Rabbit
Immunogen A synthetic peptide of human REEP2 (aa 161-174).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51308
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.