missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Regenerated Cellulose Tangential Flow Filtration Cassette for up to 0.5 L
Shop All Sartorius ProductsBeskrivning
- Link the cassettes quickly and securely to a suitable pump and reservoirs, right out of the box.
- All-in-one design avoids the need for expensive cassette holders and TFF systems.
- Advanced flow path and fixed occlusion restrictor eliminate the effort that is usually needed for TFF.
- High performance PES or Hydrosart™ RC membranes ensure maximum recoveries.
- Improve safety for you and your sample with no need for flushing or cleaning.
- Cassettes contain 30 to 60% less plastic and are supplied with only the tubing you need.
- Membrane material: Regenerated cellulose
- Recommended feed volume: 100 to 500 mL
Specifikationer
Specifikationer
| Filter Type | Ultrafiltration |
| Format | Cassette |
| Material | Polycarbonate |
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 300 kDa |
| Pore Size | 0.2 μm |
| Sterility | Non-sterile |
| Type | TFF cassette |
| Volume (Metric) | 0.5 L |
| For Use With (Equipment) | Peristaltic Pump |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?