missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Regenerated Cellulose Tangential Flow Filtration Cassette for up to 1 L

Product Code. 30178724 Shop All Sartorius Products
Click to view available options
MWCO:
10 kDa
100 kDa
2 kDa
30 kDa
300 kDa
5 kDa
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 30178724

Brand: Sartorius VFS050H0100SV

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Plug-and-play tangential flow filtration cassette that is made from regenerated cellulose, for ultrafiltration and diafiltration of intermediate sample volumes.

  • Link the cassettes quickly and securely to a suitable pump and reservoirs, right out of the box.
  • All-in-one design avoids the need for expensive cassette holders and TFF systems.
  • Advanced flow path and fixed occlusion restrictor eliminate the effort that is usually needed for TFF.
  • High performance PES or Hydrosart™ RC membranes ensure maximum recoveries.
  • Improve safety for you and your sample with no need for flushing or cleaning.
  • Cassettes contain 30 to 60% less plastic and are supplied with only the tubing you need.
  • Membrane material: Regenerated cellulose
  • Recommended feed volume: 500 to 1000 mL
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Format Cassette
For Use With (Equipment) Peristaltic Pump
Material Polycarbonate
Material (Membrane) Regenerated Cellulose
MWCO 100 kDa
Sample Volume 0.5 to 1 L
Sterility Non-sterile
Type TFF cassette
Volume (Metric) 1 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.