missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RelA/NFkB p65 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56502SS
This item is not returnable.
View return policy
Description
RelA/NFkB p65 Polyclonal specifically detects RelA/NFkB p65 in Human, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| RelA/NFkB p65 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 0.2% gelatin with 0.05% Sodium Azide | |
| MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a, CRA_c) | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.5 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 10 ug/mL | |
| Q04206 | |
| RELA | |
| A synthetic peptide corresponding to amino acids within 1 and 50 of human p65 protein was used as immunogen. | |
| 0.025 mg | |
| Apoptosis, Cancer, Immune System Diseases, Immunology, Innate Immunity, Signal Transduction | |
| 5970 | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction