Learn More
RelA/NFkB p65 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56502
This item is not returnable.
View return policy
Description
RelA/NFkB p65 Polyclonal specifically detects RelA/NFkB p65 in Human, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Western Blot | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 10 ug/mL | |
| RELA | |
| A synthetic peptide corresponding to amino acids within 1 and 50 of human p65 protein was used as immunogen. | |
| IgG |
| Polyclonal | |
| MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
| Rabbit | |
| 0.1 mg |