missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Relaxin 1 Monoclonal Antibody (361513)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561285
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Relaxins are known endocrine and autocrine/paracrine hormones, belonging to the insulin gene superfamily. In the human there are three non-allelic relaxin genes, RLN1, RLN2 and RLN3. RLN1 and RLN2 share high sequence homology. This encoded protein is synthesized as a single-chain polypeptide but the active form consists of an A chain and a B chain linked by disulfide bonds; however, their exact cleavage sites have not been described. Relaxin is produced by the ovary, and targets the mammalian reproductive system to ripen the cervix, elongate the pubic symphysis and inhibit uterine contraction. It may have additional roles in enhancing sperm motility, regulating blood pressure, controlling heart rate and releasing oxytocin and vasopressin. This gene has multiple polyadenylation sites. There are multiple alternatively spliced transcript variants described for this gene but their full length nature is not known yet.
Specifications
| Relaxin 1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P04808 | |
| Rln1 | |
| E. coli-derived recombinant human Relaxin-1, Lys26-Cys185. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Neutralization, Western Blot | |
| 361513 | |
| Unconjugated | |
| Rln1 | |
| bA12D24.3.1; bA12D24.3.2; H1; H1RLX; preprorelaxin; preprorelaxin H1; prorelaxin 1; Prorelaxin H1; RELAX; relaxin 1; Relaxin 1 (H1); Relaxin A chain; Relaxin B chain; relaxin RLXH1; Rln; Rln1; rlx; RLX bA12D24.3.1; RLXH1 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 6013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction