Learn More
Invitrogen™ RelB Recombinant Rabbit Monoclonal Antibody (24GB5070)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557684
 View more versions of this product
Product Code. 30275734
  • 564.00 € / 100µL
Description
RelB Recombinant Monoclonal Antibody for Western Blot, Flow
The NFkB transcription factor was originally identified as a protein complex consisting of a DNA-binding subunit and an associated protein. The DNAbinding subunit is functionally related to c-Rel p75 and Rel B p68. The p50 subunit was initially believed to be a functionally unique protein derived from the amino-terminus of a precursor designated p105. A second protein designated p52 (previously referred to as p49) has been identified that can act as an alternative NFkB subunit. Rel B does not bind with high affinity to NFkB sites, but heterodimers between Rel B and p50 bind with an affinity comparable to that of p50 NFkB homodimers. However, Rel B/p50 heterodimers, in contrast to NFkB heterodimers, transactivates transcription of promotors containing kB binding sites.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RelB
Recombinant Monoclonal
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q01201, Q04863
RELB
A synthesized peptide derived from human Rel B (aa 565-579).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5070
Unconjugated
RELB
avian reticuloendotheliosis viral (v-rel) oncogene related B; IREL; I-REL; OTTHUMP00000221313; rel b; RELB; REL-B; RELB proto-oncogene, NF-kB subunit; shep; transcription factor RelB; v-rel avian reticuloendotheliosis viral oncogene homolog B; v-rel avian reticuloendotheliosis viral oncogene homolog B (nuclear factor of kappa light polypeptide gene enhancer in B-cells 3); v-rel reticuloen; v-rel reticuloendotheliosis viral oncogene homolog B; v-rel reticuloendotheliosis viral oncogene homolog B, nuclear factor of kappa light polypeptide gene enhancer in B-cells 3
Rabbit
Affinity chromatography
RUO
100360982, 19698, 5971
-20°C