Learn More
Unity Lab Services™ Remote Monitoring Requalification Service
Product Code. 30178679
Change view
Click to view available options
For Use With (Equipment):
Remote Monitoring Equipment
Unit Size:
1 unit
Product Code. For Use With (Equipment) Unit Size
30178679 Remote Monitoring Equipment 1 unit
1 options
Product Code. 30178679 Supplier Unity Lab Services™ Supplier No. RQRM
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Requalification Services for Remote Monitoring (documentation included)
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications and conforms to standards and requirements.
  • Key tests include sensors and any accessories are correctly assembled and installed, and audit trails are reviewable and printable.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Remote Monitoring Equipment
Product Type Requalification Service