missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Renin Antibody (SR1820), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22259-100ul
This item is not returnable.
View return policy
Description
Renin Monoclonal antibody specifically detects Renin in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| Renin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| SR1820 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, Prorenin, renin, renin precursor, renal | |
| A synthesized peptide derived from human Renin (Uniprot #: P00797) | |
| 100 μg | |
| Cardiovascular Biology, metabolism | |
| 5972 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction