missing translation for 'onlineSavingsMsg'
Learn More

Repaglinide, MedChemExpress

Product Code. p-200097473
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16406676 100 mg 100mg
16402487 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16406676 Supplier Medchem Express Supplier No. HY15209

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Repaglinide is an insulin secretagogue for the treatment of type-2 diabetes mellitus.

Repaglinide is an insulin secretagogue for the treatment of type-2 diabetes mellitus.

Specifications

Chemical Name or Material Repaglinide
CAS 135062-02-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H36N2O4
Quantity 100 mg
Synonym AG-EE 623ZW
Solubility Information DMSO : ≥ 50 mg/mL (110.48 mM)
SMILES CC(C)C[C@H](NC(CC1=CC(OCC)=C(C(O)=O)C=C1)=O)C2=CC=CC=C2N3CCCCC3
Molecular Weight (g/mol) 452.59
Formula Weight 452.59
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.