Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.
Brand: Lovibond™ 513130BT
Code : NEW
Additional Details : CAS Number : 5329-14-6 Weight : 0.50000kg Transport : UN number : 2967 Chem class : 8 Pack group : III
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
White | |
1.9 | |
Water analysis | |
100/Pk. | |
20°C | |
Silica No. 1 Tablet |
Sodium Bisulfate 25 to 35%, Boric Acid 45 to 50%, Ammonium Heptamolybdate Tetrahydrate 10 to 20% | |
Tablet | |
Colorimeter and spectrophotometer | |
0.05 to 4mg/L | |
Silicomolybdate |
Safety and Handling
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sulfamidic acidsodium molybdate