missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RERG Polyclonal Antibody
Description
Peptide sequence: CAFYECSACT GEGNITEIFY ELCREVRRRR MVQGKTRRRS STTHVKQAIN Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 100%.
RERG, a member of the RAS superfamily of GTPases, inhibits cell proliferation and tumor formation.
Specifications
Specifications
| Antigen | RERG |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | RERG |
| Gene Accession No. | Q96A58 |
| Gene Alias | MGC15754; RAS like estrogen regulated growth inhibitor; RAS-like; RAS-like, estrogen-regulated, growth inhibitor; RAS-like, estrogen-regulated, growth-inhibitor; ras-related and estrogen-regulated growth inhibitor; ras-related and estrogen-regulated growth protein; RERG; RGD1562829; si:cabz01085950.1 |
| Gene Symbols | RERG |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?