missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical R/0150/48

Code : 21

Additional Details : CAS Number : 108-46-3 Weight : 0.10000kg Transport : UN number : 2876 Chem class : 6.1 Pack group : III


Quantity 100g

 View more versions of this product

Product Code. 10000350

Quantity Price
1 27.43 € / 100g
4 25.93 € / 100g
In stock, ready to ship: 7

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Resorcinol, Extra Pure, SLR

Specifications

Chemical Identifiers

108-46-3
110.112
GHMLBKRAJCXXBS-UHFFFAOYSA-N
5054
benzene-1,3-diol
C6H6O2
2269
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
CHEBI:27810
C1=CC(=CC(=C1)O)O

Specifications

Glass Jar
Beige
100g
108-46-3
2269
GHMLBKRAJCXXBS-UHFFFAOYSA-N
benzene-1,3-diol
5054
110.11g/mol
281°C
109°C
4.4
C6H6O2
resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
C1=CC(=CC(=C1)O)O
110.112
CHEBI:27810
Solid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Resorcinol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates