missing translation for 'onlineSavingsMsg'
Learn More

Resorcinol monobenzoate, 94%

Product Code. 11489436
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489436 500 g 500g
11479436 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489436 Supplier Thermo Scientific Alfa Aesar Supplier No. A19150.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 136-36-7 | C13H10O3 | 214.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-36-7
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00020118
InChI Key GDESWOTWNNGOMW-UHFFFAOYSA-N
Synonym resorcinol monobenzoate, 1,3-benzenediol, monobenzoate, eastman inhibitor rmb, resorcinol, monobenzoate, benzoic acid, m-hydroxyphenyl ester, 3-benzoyloxyphenol, benzoic acid 3-hydroxyphenyl ester, unii-942e82kuwd, 3-hydroxyphenyl benzoate, 1,3-benzenediol, 1-benzoate
PubChem CID 8690
IUPAC Name (3-hydroxyphenyl) benzoate
SMILES OC1=CC=CC(OC(=O)C2=CC=CC=C2)=C1

Specifications

Melting Point 130°C to 133°C
Quantity 500 g
Formula Weight 214.22
Percent Purity 94%
Chemical Name or Material Resorcinol monobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.