Learn More
Moldex™ Smart Disposable Respirator FFP3S
Product Code. 15289343
Change view
Click to view available options
Design Type:
Cup/Molded
Unit Size:
Pack of 10
Product Code. Design Type Unit Size
15289343 Cup/Molded Pack of 10
1 options
Product Code. 15289343 Supplier Moldex™ Supplier No. 250501
  • ONLINE EXCLUSIVE PRICE
    2196.91 €
    78.73 € / Pack of 10
    Save 2118.18 €
    96% Off
Estimated Shipment: 08-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Smart Disposable Masks are easy to wear and sit securely and comfortably on the skin at all times, without the need for a nose clip. Our DuraMesh™ structure preserves the shape of the mask under even the most testing conditions.
  • FFP3 NR D masks
  • Single use respirator face mask with valve
  • Reliable protection against particles
  • Soft nose cushion for enhanced wearing comfort
  • ActivForm™ 3D design for a comfortable and secure fit
  • Ventex™-Valve
  • Adjustable headstrap with clip - easy to put on and take off, or wear around the neck
Specifications
Certifications/Compliance (Europe) FFP3 NR D, EN 149:2001+A1:2009, (EU)2016/425
Design Type Cup/Molded
Disposable Single-use
Filter Type FFP3
Material Polypropylene, EVA
With Exhalation Valve Yes
Filtration Layer Material Polypropylene
For Use With (Equipment) Harmful and carcinogenic dusts, water and oil based mists/aerosols, biological agents of risk group 2 and 3, CMR-substances
Material Headstrap Polyester, Lycra
Nose Piece Material Polyethylene
Nosepiece Type Flexible nosepiece
Strap Attachment Type 1-Strap with adjustable head harness