Learn More
Resveratrol, 98%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J60790.ME
| Quantity | 500mg |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as a natural phenolic antioxidant that is a potent activator of SIRT1 and inhibitor of Cox-1. Resveratrol has also been used extensively in stem cell studies to enhance self-renewal and multipotency of mesencymal stem cel
Specifications
| 501-36-0 | |
| MFCD00133799 | |
| 14,8158 | |
| Soluble in water (3mg/100mL),ethanol (50mg/mL),DMSO (?16mg/mL),DMF (∽65mg/mL),PBS (pH 7.2) (∽100μg/mL),methanol,and acetone (50mg/mL). | |
| C1=CC(=CC=C1C=CC2=CC(=CC(=C2)O)O)O | |
| 228.247 | |
| CHEBI:45713 | |
| 98% |
| C14H12O3 | |
| 500mg | |
| resveratrol, trans-resveratrol, 3,4',5-trihydroxystilbene, 3,5,4'-trihydroxystilbene, e-resveratrol, 3,4',5-stilbenetriol, e-5-4-hydroxystyryl benzene-1,3-diol, resvida, 3,4',5-trihydroxy-trans-stilbene, 5-e-2-4-hydroxyphenyl ethenyl benzene-1,3-diol | |
| LUKBXSAWLPMMSZ-OWOJBTEDSA-N | |
| 5-[(E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol | |
| 445154 | |
| 228.24 | |
| Resveratrol |
Safety and Handling
- Resveratrol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Resveratrol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.