missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Resveratrol, 99%

Product Code. 10719098 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10719098 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10719098 Supplier Thermo Scientific Chemicals Supplier No. 430075000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Resveratrol, 3,4',5-Trihydroxystilbene, CAS # 501-36-0, is a polyphenol antioxidant found in plant foods.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description


• Resveratrol is a phytoalexin, an endogenous antiparasitic in plants
• Resveratrol is a potent antioxidant; it is found in plant foods such as grapes, blueberries and peanuts
• Resveratrol has a possible role as an NFKb inhibitor, whereby it contributes to cell survival
Applications


• Resveratrol has anti-inflammatory effects via cyclooxygenase inhibition and may also be capable of inhibiting oncogenes
• Useful for the study of cellular redox states and aging

TRUSTED_SUSTAINABILITY

Specifications

CAS 501-36-0
Melting Point 261.0°C to 263.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C14H12O3
MDL Number MFCD00133799
Quantity 500 mg
Merck Index 15,8279
Synonym resveratrol, trans-resveratrol, 3,4',5-trihydroxystilbene, 3,5,4'-trihydroxystilbene, e-resveratrol, 3,4',5-stilbenetriol, e-5-4-hydroxystyryl benzene-1,3-diol, resvida, 3,4',5-trihydroxy-trans-stilbene, 5-e-2-4-hydroxyphenyl ethenyl benzene-1,3-diol
InChI Key LUKBXSAWLPMMSZ-OWOJBTEDSA-N
SMILES C1=CC(=CC=C1C=CC2=CC(=CC(=C2)O)O)O
IUPAC Name 5-[(E)-2-(4-hydroxyphenyl)ethenyl]benzene-1,3-diol
Molecular Weight (g/mol) 228.24
PubChem CID 445154
ChEBI CHEBI:45713
Formula Weight 228.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Resveratrol
Show More Show Less
compliance-icons
Product Identifier
  • Resveratrol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.